Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Metadherin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7180
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7180 Supplier R&D Systems Supplier No. MAB7180
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

LYRIC Monoclonal specifically detects LYRIC in Mouse samples. It is validated for Western Blot.

Specifications

Antigen LYRIC
Applications Western Blot
Classification Monoclonal
Clone 754523
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q80WJ7
Gene Alias 3D3, AEG1, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRIC, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, MTDH, protein LYRIC
Gene Symbols MTDH
Host Species Rat
Immunogen E. coli-derived recombinant mouse Metadherin Lys167-Ser297 Accession # Q80WJ7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92140
Test Specificity Detects mouse Metadherin in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.