Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Meteorin Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3475
This item is not returnable.
View return policy
Description
Meteorin Monoclonal specifically detects Meteorin in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Meteorin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q8C1Q4 | |
Metrn | |
Mouse myeloma cell line NS0-derived recombinant mouse Meteorin isoform 1 Gly22-Asp291 Accession # Q8C1Q4 | |
100 μg | |
Primary | |
Detects mouse Meteorin in direct ELISAs and Western blots. | |
Mouse | |
Purified |
Western Blot, Immunocytochemistry | |
347505 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C16orf23, c380A1.2, METRN | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
79006 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction