Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Meteorin Biotinylated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems BAM34751
This item is not returnable.
View return policy
Description
Meteorin Monoclonal specifically detects Meteorin in Mouse samples. It is validated for ELISA Detection (Matched Antibody Pair).Specifications
Meteorin | |
Monoclonal | |
LYOPH | |
ELISA Detection (Matched Antibody Pair) 0.5-2 ug/mL | |
Q8C1Q4 | |
Metrn | |
Mouse myeloma cell line NS0-derived recombinant mouse Meteorin Gly22-Asp291 Accession # Q8C1Q4 | |
250 μg | |
Primary | |
Detects mouse Meteorin in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse CDNF, recombinant rat (rr) CNTF, or rrGDNF is observed. | |
Mouse | |
Purified |
ELISA | |
347504 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
C16orf23, c380A1.2, METRN | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
79006 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction