Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Meteorin-like/METRNL Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6679
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6679 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6679 Supplier R&D Systems Supplier No. AF6679
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Meteorin-like/METRNL Polyclonal specifically detects Meteorin-like/METRNL in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen Meteorin-like/METRNL
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q8VE43
Gene Alias Meteorin like, meteorin, glial cell differentiation regulator-like, meteorin-like protein, METRNL, MGC99788
Gene Symbols METRNL
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Meteorin-like/METRNL Accession # Q8VE43
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284207
Test Specificity Detects mouse Meteorin-like/METRNL in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse Meteorin is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.