Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Meteorin-like/METRNL APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC6679A025
This item is not returnable.
View return policy
Description
Meteorin-like/METRNL Monoclonal specifically detects Meteorin-like/METRNL in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Meteorin-like/METRNL | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Meteorin like, meteorin, glial cell differentiation regulator-like, meteorin-like protein, METRNL, MGC99788 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
284207 | |
Mouse | |
Supernatant |
Flow Cytometry | |
829535 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Q8VE43 | |
METRNL | |
Chinese hamster ovary cell line CHO-S-derived recombinant mouse Meteorin-like/METRNL Gln46-Glu311 Accession # Q8VE43 | |
25 Tests | |
Primary | |
Detects mouse Meteorin-like/METRNL in direct ELISA. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction