Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MFRP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3445
This item is not returnable.
View return policy
Description
MFRP Polyclonal specifically detects MFRP in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| MFRP | |
| Polyclonal | |
| Unconjugated | |
| Q8K480 | |
| MFRP | |
| Mouse myeloma cell line NS0-derived recombinant mouse MFRP Gln91-Pro584 Accession # Q8K480 | |
| 100 μg | |
| Primary | |
| Detects mouse MFRP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) MFRP is observed, 15% cross-reactivity with recombinant mouse (rm) Frizzled-4 is observed, 10% cross-reactivity with rmFrizzled-1, rhFrizzled-5, rmFrizzled-8, and rmFrizzled-10 is observed, and 5% cross-reactivity with rmFrizzled-3, rmFrizzled-6, and rmFrizzled-7 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| MCOP5, membrane frizzled-related protein, Membrane-type frizzled-related protein, MGC32938, NNO2, RD6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 83552 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction