Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MGL1/CD301a Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4938SP
This item is not returnable.
View return policy
Description
MGL1/CD301a Polyclonal specifically detects MGL1/CD301a in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
MGL1/CD301a | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Clec10a | |
Mouse myeloma cell line NS0-derived recombinant mouse MGL1/CD301a Gln57-Ser304 Accession # AAH14811 | |
25 μg | |
Primary | |
Detects mouse MGL1/CD301a in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse MGL2 is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
AAH14811 | |
Goat | |
Affinity Purified | |
RUO | |
10462 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction