Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MGMT Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3299SP
This item is not returnable.
View return policy
Description
MGMT Monoclonal specifically detects MGMT in Mouse samples. It is validated for Western Blot.Specifications
MGMT | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P26187 | |
MGMT | |
E. coli-derived recombinant mouse MGMT Met1-Asn211 Accession # P26187 | |
25 μg | |
Primary | |
Detects mouse MGMT in Western blots. | |
Mouse | |
Purified |
Western Blot | |
300008 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4255 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction