Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse MICL/CLEC12A Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB9550N100U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB9550N100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB9550N100U Supplier R&D Systems Supplier No. FAB9550N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

MICL/CLEC12A Monoclonal specifically detects MICL/CLEC12A in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen MICL/CLEC12A
Applications Flow Cytometry
Classification Monoclonal
Clone 812303
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Gene Accession No. Q504P2
Gene Alias CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MICL/CLEC12A with an N-terminal 9-His tag, Thr67-Arg267, Accession No. Q504P2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 160364.0
Test Specificity Detects mouse MICL/CLEC12A in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.