Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Midkine Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7769SP
This item is not returnable.
View return policy
Description
Midkine Polyclonal specifically detects Midkine in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Midkine | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Amphiregulin-associated protein, ARAP, MDK, MEK, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
Sheep | |
Affinity Purified | |
RUO | |
4192 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL | |
P12025 | |
MDK | |
E. coli-derived recombinant mouse Midkine Gly89-Asp140 Accession # P12025 | |
25 μg | |
Primary | |
Detects mouse Midkine in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human Midkine is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction