Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MMP-12 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3467SP
This item is not returnable.
View return policy
Description
MMP-12 Polyclonal specifically detects MMP-12 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
MMP-12 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP12, MMP-12 | |
Goat | |
Affinity Purified | |
RUO | |
4321 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
EDL24933 | |
MMP12 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMP-12 Ala18-Cys462 Accession # EDL24933 | |
25 μg | |
Primary | |
Detects mouse MMP-12 in direct ELISAs and Western blots. In direct ELISAs,less than 20% cross-reactivity with recombinant human MMP-12 is observed, and less than 5% cross-reactivity with recombinant mouse (rm) MMP-3, rmMMP-8, and rmMMP-10 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction