Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse MMP-17 ELISA Kit (Colorimetric)
SDP

Catalog No. NB180098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180098 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180098 Supplier Novus Biologicals™ Supplier No. NBP306889
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.09 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Plasma, Serum
Specificity This kit recognizes Mouse MMP-17 in samples. No significant cross-reactivity or interference between Mouse MMP-17 and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 4326
Gene Symbol MMP17
Quantity 1 Kit
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.