Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MMP-7 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC7170U100UG
This item is not returnable.
View return policy
Description
MMP-7 Monoclonal specifically detects MMP-7 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
MMP-7 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q10738 | |
MMP7 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMP-7 Met1-Leu264 Accession # Q10738 | |
RUO | |
4316 | |
Mouse | |
IgG2b |
Flow Cytometry | |
704202 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
Rat | |
100 μg | |
Primary | |
Detects mouse MMP-7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human MMP-7, recombinant mouse (rm) MMP-3, or rmMMP-8 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction