Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MMP-7 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7170SP
This item is not returnable.
View return policy
Description
MMP-7 Monoclonal specifically detects MMP-7 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
MMP-7 | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q10738 | |
MMP7 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMP-7 Met1-Leu264 Accession # Q10738 | |
25 μg | |
Primary | |
Detects mouse MMP-7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human MMP-7, recombinant mouse (rm) MMP-3, or rmMMP-8 is observed. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
704202 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4316 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction