Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MMR/CD206 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB25351G100UG
This item is not returnable.
View return policy
Description
MMR/CD206/Mannose Receptor Monoclonal specifically detects MMR/CD206/Mannose Receptor in Mouse samples. It is validated for Flow Cytometry.Specifications
MMR/CD206/Mannose Receptor | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q61830 | |
MRC1 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMR Leu19-Ala1388 Accession # Q61830 | |
RUO | |
4360 | |
Mouse | |
IgG1 |
Flow Cytometry | |
857615 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD206, CLEC13D, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen, MRC1 | |
Rat | |
100 μg | |
Primary | |
Detects mouse MMR in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction