Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MMR/CD206 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $514.50
Specifications
Antigen | MMR/CD206/Mannose Receptor |
---|---|
Clone | 310301 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
MMR/CD206/Mannose Receptor Monoclonal specifically detects MMR/CD206/Mannose Receptor in Mouse samples. It is validated for Western Blot.Specifications
MMR/CD206/Mannose Receptor | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
NP_032651 | |
4360 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMR Leu19-Ala1388 Accession # NP_032651 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse MMR in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant human MMR. |
310301 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD206, CLEC13D, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen, MRC1 | |
MRC1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title