Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse MMR/CD206 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2535P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2535P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2535P Supplier R&D Systems Supplier No. FAB2535P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 7 publications

MMR/CD206/Mannose Receptor Polyclonal specifically detects MMR/CD206/Mannose Receptor in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen MMR/CD206/Mannose Receptor
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q2HZ94
Gene Alias CD206, CLEC13D, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen, MRC1
Gene Symbols MRC1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MMR/CD206 Leu19-Ala1388 Accession # Q2HZ94
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4360
Test Specificity Detects mouse MMR/CD206 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human MMR is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.