Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse MOG Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2439
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2439 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2439 Supplier R&D Systems Supplier No. AF2439
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

MOG Polyclonal specifically detects MOG in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MOG
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q61885
Gene Alias MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein
Gene Symbols MOG
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MOG Gly29-Gly153 Accession # Q61885
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4340
Test Specificity Detects mouse MOG in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant human MOG is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.