Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Mrc2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4789SP
This item is not returnable.
View return policy
Description
Mrc2 Polyclonal specifically detects Mrc2 in Mouse samples. It is validated for Western Blot.Specifications
Mrc2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD280, CD280 antigen, CD280C-type mannose receptor 2, CLEC13E, CLEC13EUPAR-associated protein, Endo180, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, uPARAP, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein | |
Sheep | |
Affinity Purified | |
RUO | |
9902 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
AAI16643 | |
MRC2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Mrc2 Ala31-Ala1413 Accession # AAI16643 | |
25 μg | |
Primary | |
Detects mouse Mrc2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human Mrc2 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction