Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MSP/MST1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6244
This item is not returnable.
View return policy
Description
MSP/MST1 Polyclonal specifically detects MSP/MST1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MSP/MST1 | |
Polyclonal | |
Unconjugated | |
NP_032269 | |
MST1 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse MSP/MST1 Gly19-Gly716 (Cys677Ala) Accession # NP_032269 | |
100 μg | |
Primary | |
Detects mouse MSP/MST1 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human MSP is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFL, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, MST1, NF15S2, SF2 | |
Sheep | |
Affinity Purified | |
RUO | |
4485 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction