Learn More
Description
Specifications
Specifications
| Assay Range | 0.156 to 10 ng/mL |
| Assay Sensitivity | 0.094 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Gene ID (Entrez) | 4586 |
| Gene Symbol | MUC5AC |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Research Discipline | Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
