Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MyBPC3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7199SP
This item is not returnable.
View return policy
Description
MyBPC3 Monoclonal specifically detects MyBPC3 in Mouse samples. It is validated for Western Blot.Specifications
MyBPC3 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
O70468 | |
MYBPC3 | |
E. coli-derived recombinant mouse MyBPC3 Pro2-Ser169 Accession # O70468 | |
25 μg | |
Primary | |
Detects mouse MyBPC3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human MyBPC3 is observed. | |
Mouse | |
Purified |
Western Blot | |
751631 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cardiac MyBP-C, CMH4, C-protein, cardiac muscle isoform, DKFZp779E1762, FHC, MYBP-C, myosin binding protein C, cardiac, myosin-binding protein C, cardiac, myosin-binding protein C, cardiac-type | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4607 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction