Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NAC1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $503.00
Specifications
Antigen | NAC1 |
---|---|
Clone | 1243A |
Dilution | Western Blot 0.05 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
NAC1 Monoclonal specifically detects NAC1 in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
NAC1 | |
Western Blot 0.05 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, BTBD30, NAC1BEN domain containing 8, NAC-1FLJ37383, NACC1, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1 | |
Nacc1 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1243A | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Mouse | |
Q7TS78 | |
112939 | |
E. coli-derived recombinant mouse NAC1 Val457-Gln514 Accession # Q7TS78 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse NAC1 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title