Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse NAIP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7608
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7608 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7608 Supplier R&D Systems Supplier No. AF7608
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

NAIP Polyclonal specifically detects NAIP in Mouse samples. It is validated for Western Blot.

Specifications

Antigen NAIP
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9QWK5
Gene Alias baculoviral IAP repeat-containing 1, BIRC1, BIRC1baculoviral IAP repeat-containing protein 1, FLJ18088, FLJ42520, FLJ58811, Neuronal apoptosis inhibitory protein, NLR family, apoptosis inhibitory protein, NLR family, BIR domain containing 1, NLRB1, nucleotide-binding oligomerization domain, leucine rich repeat and BIR domaincontaining 1, psi neuronal apoptosis inhibitory protein, psiNAIP
Gene Symbols NAIP
Host Species Sheep
Immunogen E. coli-derived recombinant mouse NAIP Lys783-Ser856 Accession # Q9QWK5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4671
Test Specificity Detects mouse NAIP in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse NAIP2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.