Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NCAM-1/CD56 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB7820S100UG
This item is not returnable.
View return policy
Description
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Mouse samples. It is validated for Flow Cytometry.Specifications
NCAM-1/CD56 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P13595 | |
NCAM1 | |
Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56 Leu20-Thr711 Accession # P13595 | |
RUO | |
4684 | |
Mouse | |
IgG2a |
Flow Cytometry | |
809220 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD56, CD56/ NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
Rat | |
100 μg | |
Primary | |
Detects mouse NCAM-1/CD56 in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction