Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Nectin-2/CD112 PE-conjugated Antibody, R&D Systems™



Rat Monoclonal Antibody
Supplier: R&D Systems FAB3869P
This item is not returnable.
View return policy
Description
Nectin-2/CD112 Monoclonal specifically detects Nectin-2/CD112 in Mouse samples. It is validated for Flow Cytometry.Specifications
Nectin-2/CD112 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HVEB, HVEBpoliovirus receptor-like 2, MPH, nectin 2, Nectin2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2, PRR2nectin-2, PVRL2, PVRR2poliovirus receptor related 2 | |
Rat | |
Affinity Purified | |
RUO | |
5819 | |
Mouse | |
IgG2a |
Flow Cytometry | |
829038 | |
Flow Cytometry 10 uL/10^6 cells | |
P32507 | |
PVRL2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Nectin-2/CD112 Gln32-Gly351 (predicted) Accession # P32507 | |
100 Tests | |
Primary | |
Detects mouse Nectin-2/CD112 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Nectin-2 or recombinant mouse CD155/PVR is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction