Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Nectin-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Nectin-3/PVRL3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Nectin-3/PVRL3 Polyclonal specifically detects Nectin-3/PVRL3 in Mouse samples. It is validated for Western Blot.Specifications
Nectin-3/PVRL3 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CD113, CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, Nectin3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRL3, PVRR3 | |
PVRL3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9JLB9.1 | |
25945 | |
Mouse myeloma cell line NS0-derived recombinant mouse Nectin-3 Leu56-Asp400 Accession # Q9JLB9.1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Nectin-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human Nectin-3 is observed and less than 2% cross-reactivity with recombinant mouse (rm) Nectin-1 and rmNectin-4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title