Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Nectin-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Nectin-4/PVRL4 |
---|---|
Clone | 356704 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Nectin-4/PVRL4 Monoclonal specifically detects Nectin-4/PVRL4 in Mouse samples. It is validated for Western Blot.Specifications
Nectin-4/PVRL4 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q8R007 | |
81607 | |
Mouse myeloma cell line NS0-derived recombinant mouse Nectin-4 Tyr28-Ile349 Accession # Q8R007 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Nectin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human Nectin-4, 10% cross-reactivity with recombinant mouse (rm) Nectin-3, and no cross-reactivity with rmNectin-1 or rmNectin-2 is observed. |
356704 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EDSS1, Ig superfamily receptor LNIR, LNIR, LNIRnectin 4, nectin-4, Nectin4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4, PRR4DKFZp686K05193, PVRL4 | |
PVRL4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title