Learn More
Stoelting™ Mouse and Neonatal Rat Adaptor
SDP
Designed for use with traditional U-framed stereotaxic instruments like the Classic Lab Standard series
$226.50 - $3030.00
Specifications
For Use With (Application) Neuroscience
Animal Rat
View More Specs
Products 4
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-152
SDP
View Documents
Stoelting™
51625
Mouse and Neonatal Rat Adaptors
Each for $3,030.00
Only null left
Add to Cart
10-000-155
SDP
View Documents
Stoelting™
51629
Snout Clamp for Mouse Stereotaxics
Each for $618.00
Only null left
Add to Cart
10-000-171
SDP
View Documents
Stoelting™
51648
Stereotaxic Ear Bars/Rubber Tips
Pair for $226.50
Only null left
Add to Cart
10-000-176
SDP
View Documents
Stoelting™
51655
Mouse Posts, Neonatal Rat Adaptors
Pack of 3 for $1,409.00
Only null left
Add to Cart
Description
Description
Model organisms with soft skulls, such as the mouse and the neonatal rat, are difficult to secure firmly in a traditional stereotaxic instrument. The stainless steel ear bars used for adult rats are too heavy to provide a secure hold without compressing the sides of the skull inward (which closes the airways and impedes blood fl ow). Further, the neonatal rat pup has no external opening of the auditory canal into which to insert the ear bars.
  • Ear and bite bar heights can be independently adjusted to accommodate a wide range of ages and assist in leveling the skull
  • Employs light, Delrin™ ear bars with tapered points on one end, and rubber tips on the other, to facilitate surgery on mice and rat pups
  • Unique feature permits ear bars to be independently adjusted in height to level the skull
  • Laser engraved scales show the vertical positions of the ear bars
  • Tooth bar and nose clamp secures the nose
  • Length accommodates larger, more mature animals
  • Specialized jaw holder cuffs securely clamp the zygomatic processes of the skull
  • Jaw holder cuffs fit over the ends of the ear bars, replacing the rubber tips, providing an alternative non-invasive means of securing the animal's head in the stereotaxic stage
  • Options for triple point securing of the animal's head in this adaptor make it exceptionally versatile
  • Set of two jaw holder cuffs included
Specifications
Specifications
Neuroscience
Rat
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.