Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Nephronectin Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $561.50
Specifications
Antigen | Nephronectin |
---|---|
Concentration | LYOPH |
Dilution | Immunohistochemistry 1-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Description
Nephronectin Polyclonal specifically detects Nephronectin in Mouse samples. It is validated for Immunohistochemistry.Specifications
Nephronectin | |
Immunohistochemistry 1-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
EGFL6L, Nctn, nephronectin, NPNT, POEM | |
NPNT | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
AAH68308 | |
255743 | |
Mouse myeloma cell line NS0-derived recombinant mouse Nephronectin Asp20-Cys561 Accession # AAH68308 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Nephronectin in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title