Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Netrin-4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF1132
This item is not returnable.
View return policy
Description
Netrin-4 Polyclonal specifically detects Netrin-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
Netrin-4 | |
Polyclonal | |
Unconjugated | |
Q9JI33 | |
Ntn4 | |
Mouse myeloma cell line NS0-derived recombinant mouse Netrin-4 Leu19-Val628 Accession # Q9JI33 | |
100 μg | |
Primary | |
Detects mouse Netrin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximatley 25% cross-reactivity with recombinant human Netrin-4 and less than 1% cross-reactivity with recombinant mouse Netrin-1, recombinant chicken Netrin-2, and recombinant mouse Netrin-G1a. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
beta-netrin, hepar-derived netrin-like protein, netrin 4, Netrin4, NTN4, PRO3091 | |
Goat | |
Affinity Purified | |
RUO | |
59277 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction