Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Netrin-G1a Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1166
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1166 500 μg
MAB1166SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1166 Supplier R&D Systems Supplier No. MAB1166
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Netrin-G1a Monoclonal specifically detects Netrin-G1a in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Netrin-G1a
Applications Western Blot
Classification Monoclonal
Clone 229722
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8R4G0
Gene Alias Lmnt1, NetrinG1a, NTG1, NTNG1
Gene Symbols Ntng1
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Netrin-G1a His29-Gly513 Accession # Q8R4G0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22854
Test Specificity Detects mouse Netrin-G1a in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant chicken (rch) Netrin-1, recombinant mouse Netrin-1, rchNetrin-2, or recombinant human Netrin-4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.