Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Netrin-G2a Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Netrin-G2a |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Netrin-G2a Polyclonal specifically detects Netrin-G2a in Mouse samples. It is validated for Western Blot.Specifications
Netrin-G2a | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
84628 | |
Mouse myeloma cell line NS0-derived recombinant mouse Netrin-G2a Asp18-Arg504 Accession # NP_598007 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Netrin-G2a in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Netrin-1, rmNetrin-G1a, and rmNetrin-4 is observed and less than 1% cross-reactivity with recombinant chicken Netrin-2 is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_598007 | |
Ntng2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title