Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Neuropoietin/NP Antibody, R&D Systems™

Goat Polyclonal Antibody
$164.60 - $496.92
Specifications
| Antigen | Neuropoietin/NP |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Neuropoietin/NP Polyclonal specifically detects Neuropoietin/NP in Mouse samples. It is validated for Western Blot.Specifications
| Neuropoietin/NP | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| Cardiotrophin-2, Ctf2, CTF2P, Gm494, NP | |
| Ctf2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P83714 | |
| 100289419 | |
| E. coli-derived recombinant mouse Neuropoietin/NP Ala23-Ala204 Accession # P83714 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Neuropoietin/NP in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title