Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Neutrophil Elastase/ELA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4517
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4517 Supplier R&D Systems Supplier No. AF4517
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Neutrophil Elastase/ELA2 Polyclonal specifically detects Neutrophil Elastase/ELA2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Neutrophil Elastase/ELA2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q3UP87
Gene Alias Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Gene Symbols ELANE
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Neutrophil Elastase/ELA2 Ser27-Arg260 Accession # Q3UP87
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1991
Test Specificity Detects mouse Neutrophil Elastase/ELA2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human ELA2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.