Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
R&D Systems™ Mouse Neutrophil Elastase/ELA2 Quantikine ELISA Kit
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                1 Kit
                            
                        
                    Description
The Mouse Neutrophil Elastase/ELA2 Quantikine ELISA Kit quantifies mouse Neutrophil Elastase/ELA2 in cell culture supernates (50 pg/ml), tissue lysates (13 pg/ml), serum (10 pg/ml), edta plasma (10 pg/ml), heparin plasma (10 pg/ml).
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
                
            Specifications
Specifications
| Assay Range | 31.30 to 2000 pg/mL | 
| Assay Sensitivity | 3.37 pg/mL | 
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates | 
| Specificity | Natural and Recombinant Human GDNF | 
| Gene ID (Entrez) | 1991 | 
| Host Species | Mouse | 
| Quantity | 1 Kit | 
| Sample Volume | Cell Culture Supernates (50 μL),Tissue Lysates (13 μL),Serum (10 μL),EDTA Plasma (10 μL),Heparin Plasma (10 μL) | 
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. | 
| Synonym | Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | 
| Show More | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            