Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NFIL3/E4BP4 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC8888V100UG
This item is not returnable.
View return policy
Description
NFIL3 Monoclonal specifically detects NFIL3 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
NFIL3 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
E4BP4, E4BP4IL3BP1, IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A | |
E. coli-derived recombinant mouse NFIL3/E4BP4 Lys140-Arg462 Accession # O08750 | |
RUO | |
4783 | |
Mouse | |
IgG |
Flow Cytometry | |
1218A | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
O08750 | |
Rabbit | |
100 μg | |
Primary | |
Detects NFIL3/E4BP4 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction