Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse NFIL3/E4BP4 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC8888N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC8888N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC8888N100 Supplier R&D Systems Supplier No. IC8888N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NFIL3 Monoclonal specifically detects NFIL3 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen NFIL3
Applications Flow Cytometry
Classification Monoclonal
Clone 1218A
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O08750
Gene Alias E4BP4, E4BP4IL3BP1, IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A
Host Species Rabbit
Immunogen E. coli-derived recombinant mouse NFIL3/E4BP4 Lys140-Arg462 Accession # O08750
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4783
Test Specificity Detects NFIL3/E4BP4 in direct ELISAs.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.