Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NG2/MCSP Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB6689SP
This item is not returnable.
View return policy
Description
NG2/MCSP Monoclonal specifically detects NG2/MCSP in Mouse samples. It is validated for Immunohistochemistry.Specifications
NG2/MCSP | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q8VHY0 | |
CSPG4 | |
Mouse myeloma cell line NS0-derived recombinant mouse NG2/MCSP Leu1587-Ser2229 Accession # Q8VHY0 | |
25 μg | |
Primary | |
Detects mouse NG2/MCSP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human NG2/MCSP/CSPG4 is observed. | |
Mouse | |
Purified |
Immunohistochemistry | |
546930 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AN2, chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), CSPG4, EC 3.6.3, HMW-MAA, MCSP, MCSPChondroitin sulfate proteoglycan NG2, MCSPG, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1464 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction