Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NG2/MCSP Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB6689
This item is not returnable.
View return policy
Description
NG2/MCSP Monoclonal specifically detects NG2/MCSP in Mouse samples. It is validated for Immunohistochemistry.Specifications
NG2/MCSP | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q8VHY0 | |
CSPG4 | |
Mouse myeloma cell line NS0-derived recombinant mouse NG2/MCSP Leu1587-Ser2229 Accession # Q8VHY0 | |
100 μg | |
Primary | |
Detects mouse NG2/MCSP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human NG2/MCSP/CSPG4 is observed. | |
Mouse | |
Purified |
Immunohistochemistry | |
546930 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AN2, chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), CSPG4, EC 3.6.3, HMW-MAA, MCSP, MCSPChondroitin sulfate proteoglycan NG2, MCSPG, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1464 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction