Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Nidogen-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6760
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6760 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6760 Supplier R&D Systems Supplier No. MAB6760
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Nidogen-2 Monoclonal specifically detects Nidogen-2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Nidogen-2
Applications Western Blot
Classification Monoclonal
Clone 780611
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O88322
Gene Alias NID2, NID-2, nidogen 2 (osteonidogen), nidogen-2, Nidogen2, osteonidogen
Gene Symbols NID2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Nidogen-2 Leu31-Lys1403 Accession # O88322
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22795
Test Specificity Detects mouse Nidogen-2 in direct ELISAs. In direct ELISAs, approximately 80% cross-reactivity with recombinant human (rh) Nidogen-2 and no cross-reactivity with rhNidogen-1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.