Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NKG2C/CD159c Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB1382S100UG
This item is not returnable.
View return policy
Description
NKG2C/CD159c/KLRC2 Monoclonal specifically detects NKG2C/CD159c/KLRC2 in Mouse samples. It is validated for Flow Cytometry.Specifications
| NKG2C/CD159c/KLRC2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor | |
| Mouse NKG2C/CD159c synthetic peptide Accession # NP_034783 | |
| 100 μg | |
| 3822 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 2098A | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_034783 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects mouse NKG2C/CD159c in direct ELISAs. In Flow Cytometry, no cross-reactivity with mouse NKG2A/CD159a is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction