Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NKG2C/CD159c Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $514.50
Specifications
Antigen | NKG2C/CD159c/KLRC2 |
---|---|
Clone | 2098A |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1382SP
![]() |
R&D Systems
MAB1382SP |
25 μg |
Each for $214.00
|
|
|||||
MAB1382100
![]() |
R&D Systems
MAB1382100 |
100 μg |
Each for $514.50
|
|
|||||
Description
NKG2C/CD159c/KLRC2 Monoclonal specifically detects NKG2C/CD159c/KLRC2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
NKG2C/CD159c/KLRC2 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Mouse | |
NP_034783 | |
3822 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
2098A | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor | |
Mouse NKG2C/CD159c synthetic peptide Accession # NP_034783 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse NKG2C/CD159c in direct ELISAs. In Flow Cytometry, no cross-reactivity with mouse NKG2A/CD159a is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title