Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NKG2D/CD314 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB1547S100UG
This item is not returnable.
View return policy
Description
NKG2D/CD314 Monoclonal specifically detects NKG2D/CD314 in Mouse samples. It is validated for Flow Cytometry.Specifications
NKG2D/CD314 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O54709 | |
KLRK1 | |
Mouse myeloma cell line NS0-derived recombinant mouse NKG2D Phe94-Val232 Accession # O54709 | |
100 μg | |
Primary | |
Detects mouse NKG2D in direct ELISAs and Western blots. Does not cross-react with recombinant human NKG2D. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Flow Cytometry | |
191004 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
22914 | |
Mouse | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction