Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NKG2D/CD314 Biotinylated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAM1547
This item is not returnable.
View return policy
Description
NKG2D/CD314 Monoclonal specifically detects NKG2D/CD314 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
NKG2D/CD314 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q4FJM0 | |
KLRK1 | |
Mouse myeloma cell line NS0-derived recombinant mouse NKG2D/CD314 Phe94-Val232 Accession # Q4FJM0 | |
100 μg | |
Primary | |
Detects mouse NKG2D/CD314 in direct ELISAs and Western blots. | |
Mouse | |
Purified |
Western Blot, Immunocytochemistry | |
191004 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
22914 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction