Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse NKX3.1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6475
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6475 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6475 Supplier R&D Systems Supplier No. MAB6475
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

NKX3.1 Monoclonal specifically detects NKX3.1 in Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen NKX3.1
Applications Immunocytochemistry
Classification Monoclonal
Clone 738907
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P97436
Gene Alias BAPX2, BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3-1, NKX3A, NKX3ANK homeobox, family 3, A
Gene Symbols NKX3-1
Host Species Rat
Immunogen E. coli-derived recombinant mouse NKX3.1 Met1-Pro124 (predicted) Accession # P97436
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4824
Test Specificity Detects mouse NKX3.1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKX3.1 or rhNKX6.1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.