Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Nodal Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1315
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1315 500 μg
MAB1315SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1315 Supplier R&D Systems Supplier No. MAB1315
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Nodal Monoclonal specifically detects Nodal in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Nodal
Applications Western Blot
Classification Monoclonal
Clone 209009
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P43021
Gene Alias BMP-16, MGC138230, nodal homolog, nodal homolog (mouse), nodal, mouse, homolog
Gene Symbols NODAL
Host Species Rat
Immunogen E. coli-derived recombinant mouse Nodal His245-Leu354 Accession # P43021
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4838
Test Specificity Detects mouse Nodal in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 25 - 50% cross-reactivity with recombinant Drosophila (rd) Dpp and no cross-reactivity with rmArtemin, rmCripto, rhGDNF, rhLAP, rhLatent TGF-beta 1, rhLefty-A, rmLefty-1, rrMIS, rhNeurturin, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.