Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Nogo-B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6596
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6596 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6596 Supplier R&D Systems Supplier No. MAB6596
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Nogo-B Monoclonal specifically detects Nogo-B in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen Nogo-B
Applications Immunohistochemistry
Classification Monoclonal
Clone 523808
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99P72
Gene Symbols Rtn4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Nogo-B. Met1-Ile183 (predicted) Accession # Q99P72
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57142
Test Specificity Detects mouse Nogo-B in direct ELISAs. In direct ELISAs, less than 25% cross-reactivity with recombinant human (rh) Nogo-B and recombinant rat Nogo-A and no cross-reactivity with rhNogo-A is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.