Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Nogo Receptor/NgR Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1659
This item is not returnable.
View return policy
Description
Nogo Receptor/NgR Monoclonal specifically detects Nogo Receptor/NgR in Mouse samples. It is validated for Western Blot.Specifications
| Nogo Receptor/NgR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q99PI8 | |
| RTN4R | |
| Mouse myeloma cell line NS0-derived recombinant mouse NgR Cys27-Ser447 Accession # Q99PI8 | |
| 500 μg | |
| Primary | |
| Detects mouse NgR in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human NgR. | |
| Mouse | |
| Purified |
| Western Blot | |
| 202604 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 65078 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction