Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Nogo Receptor/NgR Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1440
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1440 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1440 Supplier R&D Systems Supplier No. BAF1440
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Nogo Receptor/NgR Polyclonal specifically detects Nogo Receptor/NgR in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Nogo Receptor/NgR
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q99PI8
Gene Alias NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R
Gene Symbols RTN4R
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Nogo Receptor/NgR Cys27-Ser447 Accession # Q99PI8
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65078
Test Specificity Detects mouse Nogo Receptor/NgR in Western blots. In this format, approximately 25% cross-reactivity with recombinant human Nogo Receptor is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.